missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3-chlorosulfonylthiophene-2-carboxylate, 95%

Product Code. 15465816
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465816 1 g 1g
15475816 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465816 Supplier Alfa Aesar™ Supplier No. H60521.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 59337-92-7
Molecular Formula C6H5ClO4S2
Molecular Weight (g/mol) 240.67
MDL Number MFCD00068160
InChI Key PJVJBDAUWILEOG-UHFFFAOYSA-N
Synonym methyl 3-chlorosulfonyl thiophene-2-carboxylate, 2-carbomethoxy-3-thiophenesulfonyl chloride, methylchlorosulfonylthiophenecarboxylate, methyl 3-chlorosulfonyl-2-thiophenecarboxylate, 3-chlorosulfonyl-2-thiophene carboxylic acid methyl ester, methyl-3-chlorosulfonylthiophene-2-carboxylate, 2-methoxycarbonyl thiophene-3-sulfonyl chloride, 2-thiophenecarboxylic acid, 3-chlorosulfonyl-, methyl ester, 2-methoxycarbonyl-3-thiophenesulfonyl chloride, 2-methoxycarbonyl thiophene-3-sulphonyl chloride
PubChem CID 2736733
IUPAC Name methyl 3-(chlorosulfonyl)thiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)S(Cl)(=O)=O

Specifikationer

Melting Point 61°C to 64°C
Quantity 1 g
UN Number UN3261
Formula Weight 240.68
Percent Purity 95%
Chemical Name or Material Methyl 3-chlorosulfonylthiophene-2-carboxylate
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.