missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3-chlorosulfonylthiophene-2-carboxylate, 95%

Product Code. 15475816
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475816 5 g 5g
15465816 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475816 Supplier Alfa Aesar™ Supplier No. H60521.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 59337-92-7
Molecular Formula C6H5ClO4S2
Molecular Weight (g/mol) 240.67
MDL Number MFCD00068160
InChI Key PJVJBDAUWILEOG-UHFFFAOYSA-N
Synonym methyl 3-chlorosulfonyl thiophene-2-carboxylate, 2-carbomethoxy-3-thiophenesulfonyl chloride, methylchlorosulfonylthiophenecarboxylate, methyl 3-chlorosulfonyl-2-thiophenecarboxylate, 3-chlorosulfonyl-2-thiophene carboxylic acid methyl ester, methyl-3-chlorosulfonylthiophene-2-carboxylate, 2-methoxycarbonyl thiophene-3-sulfonyl chloride, 2-thiophenecarboxylic acid, 3-chlorosulfonyl-, methyl ester, 2-methoxycarbonyl-3-thiophenesulfonyl chloride, 2-methoxycarbonyl thiophene-3-sulphonyl chloride
PubChem CID 2736733
IUPAC Name methyl 3-(chlorosulfonyl)thiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)S(Cl)(=O)=O

Specifications

Melting Point 61°C to 64°C
Quantity 5 g
UN Number UN3261
Formula Weight 240.68
Percent Purity 95%
Chemical Name or Material Methyl 3-chlorosulfonylthiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.