missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3-chlorothiophene-2-carboxylate, 97%

Product Code. 11429686
Change view
Click to view available options
Quantity:
1g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429686 1g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11429686 Supplier Alfa Aesar™ Supplier No. B20224.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 88105-17-3
Boiling Point 72°C to 74°C (0.2mmHg)
Molecular Formula C6H5ClO2S
MDL Number MFCD00068135
Quantity 1g
Synonym methyl3-chlorothiophene-2-carboxylate, methyl 3-chloro-2-thiophenecarboxylate, 3-chlorothiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-chloro-, methyl ester, 3-chloro-2-thiophenecarboxylic acid methyl ester, pubchem9775, acmc-20ai7j, rarechem al bf 1228, maybridge1_003830, buttpark 37\08-38
InChI Key ZWKYYONTMGVPSP-UHFFFAOYSA-N
SMILES COC(=O)C1=C(C=CS1)Cl
IUPAC Name methyl 3-chlorothiophene-2-carboxylate
Molecular Weight (g/mol) 176.614
PubChem CID 2777566
Formula Weight 176.62
Percent Purity 97%
Chemical Name or Material Methyl 3-chlorothiophene-2-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.