missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-cyclohexenecarboxylate, 97%, Thermo Scientific™

Product Code. 15259786 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15259786 100 mL 100mL
15249786 25 mL 25mL
15269786 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15259786 Supplier Thermo Scientific Acros Supplier No. 452861000
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6493-77-2
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00053266
InChI Key IPUNVLFESXFVFH-UHFFFAOYNA-N
Synonym methyl cyclohex-3-enecarboxylate, 3-cyclohexene-1-carboxylic acid methyl ester, methyl 3-cyclohexenecarboxylate, 3-cyclohexene-1-carboxylic acid, methyl ester, methyl 3-cyclohexene-1-carboxylate, 1-carbomethoxy-3-cyclohexene, methyl 4-cyclohexenecarboxylate, methyl 1,2,3,6-tetrahydrobenzoate, 1,3-butadiene-methyl acrylate adduct, acmc-1bity
PubChem CID 96926
IUPAC Name methyl cyclohex-3-ene-1-carboxylate
SMILES COC(=O)C1CCC=CC1

Specifications

Density 1.0200g/mL
Boiling Point 185.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 100 mL
Specific Gravity 1.02
Formula Weight 140.18
Percent Purity 97%
Chemical Name or Material Methyl 3-cyclohexenecarboxylate
compliance-icons
Product Identifier
  • Methyl 3-cyclohexenecarboxylate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.