missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydrazinothiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10222452
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10222452 1 g Amber Glass Bottle 1g
10475122 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10222452 Supplier Thermo Scientific Maybridge Supplier No. GK01751DA
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 75681-13-9
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00052080
InChI Key RWGYQWWIXLVGDJ-UHFFFAOYSA-N
Synonym methyl 3-hydrazinothiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-hydrazinyl-, methyl ester, 3-hydrazino-2-thiophenecarboxylic acid methyl ester, 3-hydrazinothiophene-2-carboxylic acid methyl ester, 3-hydrazino thiophene-2-carboxylic acid methyl ester
PubChem CID 2777595
IUPAC Name methyl 3-hydrazinylthiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)NN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 172.21
Percent Purity 97%
Chemical Name or Material Methyl 3-hydrazinothiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.