missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydroxythiophene-2-carboxylate, 97%

Product Code. 11449647
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449647 25 g 25g
11429647 1 g 1g
11439647 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449647 Supplier Thermo Scientific Alfa Aesar Supplier No. B22129.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5118-06-9 | C6H6O3S | 158.17 g/mol

Methyl 3-hydroxythiophene-2-carboxylate is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-hydroxythiophene-2-carboxylate is used in medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5118-06-9
MDL Number MFCD00055642

Specifications

Melting Point 44°C to 48°C
Boiling Point 107°C to 109°C (13 mmHg)
Molecular Formula C6H6O3S
Quantity 25 g
Synonym methyl 3-hydroxy-2-thiophenecarboxylate, 3-hydroxy-2-thiophenecarboxylic acid methyl ester, 3-hydroxy-thiophene-2-carboxylic acid methyl ester, 3-hydroxythiophene-2-carboxylic acid methyl ester, pubchem7744, acmc-1aqhj, maybridge3_005989, 3-hydroxy-2-methoxycarbonylthiophene, 3-hydroxy-2-methoxycarbonyl-thiophene, methyl 3-hydroxythiophen-2-caboxylate
Solubility Information Slightly soluble in water.
InChI Key FYOPNADYSPZUAT-WAYWQWQTSA-N
SMILES COC(=O)C1=C(C=CS1)O
IUPAC Name methyl 3-hydroxythiophene-2-carboxylate
Molecular Weight (g/mol) 158.17
PubChem CID 581127
Formula Weight 158.18
Percent Purity 97%
Chemical Name or Material Methyl 3-hydroxythiophene-2-carboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.