missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-isocyanatobenzoate, 97%, Maybridge™

Product Code. 10525465
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10525465 10 g Amber Glass Bottle 10g
10075752 1 g Amber Glass Bottle 1g
10471295 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10525465 Supplier Thermo Scientific Maybridge Supplier No. AC31920EA
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 41221-47-0
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
MDL Number MFCD00037077
InChI Key WBGWGERFPSYHDT-UHFFFAOYSA-N
Synonym 3-carbomethoxyphenyl isocyanate, 3-isocyanatobenzoic acid methyl ester, benzoic acid, 3-isocyanato-, methyl ester, 3-methoxycarbonyl phenyl isocyanate, acmc-20akh8, methyl-3-isocyanatobenzoate, 3-carbomethoxyphenylisocyanate, methyl 3-isocyanatobenzoate, 3-isocyanato-benzoic acid methyl ester, m-methoxycarbonylphenyl isocyanate
PubChem CID 2733366
IUPAC Name methyl 3-isocyanatobenzoate
SMILES COC(=O)C1=CC=CC(=C1)N=C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 177.16
Chemical Name or Material Methyl 3-isocyanatobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.