missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(Methyl(pentyl)amino)propanoate, TRC

Product Code. 30480038
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480038 25 mg 25mg
30478147 10 mg 10mg
30472533 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30480038 Supplier Toronto Research Chemicals Supplier No. TRCM33487125MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 744266-99-7
Molecular Formula C10H21NO2
Molecular Weight (g/mol) 187.28
Synonym Ibandronate Sodium in-House Impurity, N-Methyl-N-pentyl-beta-alanine Methyl Ester
IUPAC Name methyl 3-[methyl(pentyl)amino]propanoate
SMILES CCCCCN(C)CCC(=O)OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H21NO2/c1-4-5-6-8-11(2)9-7-10(12)13-3/h4-9H2,1-3H3
Formula Weight 187.1572
Chemical Name or Material Methyl 3-(Methyl(pentyl)amino)propanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.