missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-oxocyclobutanecarboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15448688
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448688 5 g 5g
15458688 250 mg 250mg
15438688 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448688 Supplier Thermo Scientific Alfa Aesar Supplier No. H62110.06
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Methyl 3-oxocyclobutanecarboxylate can be used as an intermediate for the synthesis of various pharmaceutical compounds. It is used for the preparation of novel imidazobenzazepine derivatives as dual H1/5-HT2A antagonists for the treatment of sleep disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-oxocyclobutanecarboxylate can be used as an intermediate for the synthesis of various pharmaceutical compounds. It is used for the preparation of novel imidazobenzazepine derivatives as dual H1/5-HT2A antagonists for the treatment of sleep disorders.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 695-95-4
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00604222
InChI Key IHLHSAIBOSSHQV-UHFFFAOYSA-N
Synonym methyl 3-oxocyclobutanecarboxylate, methyl 3-oxo-cyclobutanecarboxylate, 3-methoxycarbonylcyclobutanone, cyclobutanecarboxylic acid, 3-oxo-, methyl ester, 3-oxocyclobutanecarboxylic acid methyl ester, ksc630k4p, methyl 3-oxocyclobutane carboxylate, 3-oxo-1-cyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylicacid, 3-oxo-, methyl ester
PubChem CID 13992367
IUPAC Name methyl 3-oxocyclobutane-1-carboxylate
SMILES COC(=O)C1CC(=O)C1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 128.12
Percent Purity 97%
Chemical Name or Material Methyl 3-oxocyclobutanecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.