missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-oxotetrahydrothiophene-2-carboxylate, 98%, Thermo Scientific™

Product Code. 11325857
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11325857 5 g 5g
11315857 250 mg 250mg
11305857 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11325857 Supplier Thermo Scientific Alfa Aesar Supplier No. 043634.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 2689-69-2
Molecular Formula C6H8O3S
Molecular Weight (g/mol) 160.187
MDL Number MFCD00156156
InChI Key TUSSXVYKOOVOID-UHFFFAOYSA-N
Synonym methyl 3-oxotetrahydrothiophene-2-carboxylate, 2-thiophenecarboxylic acid, tetrahydro-3-oxo-, methyl ester, methyl tetrahydro-3-oxo-2-thenoate, methyl 3-oxo-2,3,4,5-tetrahydrothiophene-2-carboxylate, acmc-1cmcz, 2-methoxycarbonyl-3-oxotetrahydrothiophene, methyl 3-oxo-tetrahydrothiophene-2-carboxylate, 2-carbomethoxy-2,3,4,5-tetrahydrothiophen-3-one, methyl 3-oxo-2,4,5-trihydrothiophene-2-carboxylate
PubChem CID 102870
IUPAC Name methyl 3-oxothiolane-2-carboxylate
SMILES COC(=O)C1C(=O)CCS1

Spécification

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 160.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 3-oxotetrahydrothiophene-2-carboxylate
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.