missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-trifluoromethylbenzoylacetate, 95%, Thermo Scientific™

Product Code. 10341551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10341551 2.5 g Glass bottle 2.5g
10731715 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10341551 Supplier Thermo Scientific Acros Supplier No. 425450025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 93618-66-7
Molecular Formula C11H9F3O3
Molecular Weight (g/mol) 246.19
MDL Number MFCD00216522
InChI Key RPRMYRPHNDGZOY-UHFFFAOYSA-N
Synonym methyl 3-oxo-3-3-trifluoromethyl phenyl propanoate, methyl 3-trifluoromethyl benzoylacetate, methyl 3-trifluoromethyl benzoyl acetate, 3-3-trifluoromethylphenyl-3-oxo-propionic acid methyl ester, 3-oxo-3-3-trifluoromethyl-phenyl-propionic acid methyl ester, pubchem2695, methyl 3-trifluoromethylbenzoylacetate, methyl 3-oxo-3-3-trifluoromethylphenyl propanoate, 3-oxo-3-3-trifluoromethylphenyl propionic acid methyl ester
PubChem CID 735880
IUPAC Name methyl 3-oxo-3-[3-(trifluoromethyl)phenyl]propanoate
SMILES COC(=O)CC(=O)C1=CC=CC(=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 94°C to 96°C (0.2 mmHg)
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.4864 to 1.4884
Quantity 2.5 g
Solubility Information Solubility in water: neglegible in water
Formula Weight 246.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 3-trifluoromethylbenzoylacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.