missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(1H-imidazol-1-ylmethyl)benzoate, 97%, Thermo Scientific™

Product Code. 10777295
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10777295 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10777295 Supplier Thermo Scientific Maybridge Supplier No. CC43322DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 160446-18-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H12N2O2
MDL Number MFCD00608318
Quantity 1 g
Synonym methyl 4-1h-imidazol-1-ylmethyl benzoate, methyl 4-imidazol-1-ylmethyl benzoate, methyl 4-1h-imidazol-1-yl methyl benzoate, methyl 4-imidazolylmethyl benzoate, 4-imidazol-1-ylmethyl-benzoic acid methyl ester, 4-1h-imidazol-1-ylmethyl benzoic acid methyl ester, benzoicacid, 4-1h-imidazol-1-ylmethyl-, methyl ester
InChI Key HMWIIHIOSBESMH-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=C(C=C1)CN2C=CN=C2
IUPAC Name methyl 4-(imidazol-1-ylmethyl)benzoate
Molecular Weight (g/mol) 216.24
PubChem CID 3164140
Formula Weight 216.24
Percent Purity 97%
Chemical Name or Material Methyl 4-(1H-imidazol-1-ylmethyl)benzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.