missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-[3-(dimethylamino)propoxy]benzoate, 97%, Thermo Scientific™

Product Code. 10225554
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225554 1 g Amber Glass Bottle 1g
11316845 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225554 Supplier Thermo Scientific Maybridge Supplier No. CC59522DA
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 190660-97-0
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.30
MDL Number MFCD06203847
InChI Key SRXQDDFKGDLAJM-UHFFFAOYSA-N
Synonym methyl 4-3-dimethylamino propoxy benzoate, 4-3-dimethylamino-propoxy-benzoic acid methyl ester, methyl 4-3-n,n-dimethylamino-1-propyloxy benzoate, methyl-4-3-n,n-dimethylamino-1-propyloxy benzoate
PubChem CID 18508114
IUPAC Name methyl 4-[3-(dimethylamino)propoxy]benzoate
SMILES COC(=O)C1=CC=C(OCCCN(C)C)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 237.3
Percent Purity 97%
Chemical Name or Material Methyl 4-[3-(dimethylamino)propoxy]benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.