missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,5-dibromothiophene-2-carboxylate, 97%

Product Code. 15414797
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414797 5 g 5g
15404797 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414797 Supplier Thermo Scientific Alfa Aesar Supplier No. H32113.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 62224-24-2 | C6H4Br2O2S | 299.964 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62224-24-2
Molecular Formula C6H4Br2O2S
Molecular Weight (g/mol) 299.964
MDL Number MFCD00831688
InChI Key URPMSPAEAVFKCO-UHFFFAOYSA-N
Synonym methyl 4,5-dibromo-2-thiophenecarboxylate, 4,5-dibromothiophene-2-carboxylic acid methyl ester, maybridge3_005092, methyldibromothiophenecarboxylate, methyl 4,5-dibromo-2-thiophencarboxylate, methyl 4, 5-dibromothiophene-2-carboxylate, 4,5-dibromo-thiophene-2-carboxylic acid methyl ester
PubChem CID 2727707
IUPAC Name methyl 4,5-dibromothiophene-2-carboxylate
SMILES COC(=O)C1=CC(=C(S1)Br)Br

Specifications

Melting Point 77°C to 81°C
Quantity 5 g
Formula Weight 299.97
Percent Purity 97%
Chemical Name or Material Methyl 4,5-dibromothiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.