missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,5-dimethoxy-3-hydroxybenzoate, 98%, Thermo Scientific™

Product Code. 10766432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Product Code. Quantity Packaging unitSize
10766432 5 g Glass bottle 5g
1 options
This item is not returnable. View return policy

Product Code. 10766432

Brand: Thermo Scientific Acros 344540050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 83011-43-2
Melting Point 80°C to 84°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Glass bottle
Molecular Formula C10H12O5
MDL Number MFCD00598192
Quantity 5 g
Synonym 3,4-dimethoxy-5-hydroxybenzoic acid methyl ester, methyl 3,4-dimethoxy-5-hydroxybenzoate, methyl 4,5-dimethoxy-3-hydroxybenzoate, methyl 3,4-methoxy-5-hydroxybenzoate, methyl 5-hydroxy-3,4-dimethoxybenzoate, methyl 3,4-dimehtoxy-5-hydroxybenzoate, methyl 4?5-dimethoxy-3-hydroxybenzoate, 3,4-dimethoxy-5-hydroxybenzoate, methyl 3,4-dimethoxy-5-hydroxy-benzoate, methyl 3-hydroxy-4,5-dimethoxy-benzoate
InChI Key LCIFXEQPXQVBGL-UHFFFAOYSA-N
SMILES COC(=O)C1=CC(O)=C(OC)C(OC)=C1
IUPAC Name methyl 3-hydroxy-4,5-dimethoxybenzoate
Molecular Weight (g/mol) 212.2
PubChem CID 2733956
Formula Weight 212.2
Percent Purity 0.98
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl 4, 5-dimethoxy-3-hydroxybenzoate, 0.98
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4,5-dimethoxy-3-hydroxybenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.