missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-acetylbenzoate, 98%

Product Code. 10045970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10045970 1 g Glass bottle 1g
10500004 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10045970 Supplier Thermo Scientific Acros Supplier No. 424770010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3609-53-8 | C10H10O3 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3609-53-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00216474
InChI Key QNTSFZXGLAHYLC-UHFFFAOYSA-N
Synonym benzoic acid, 4-acetyl-, methyl ester, unii-3s3d53j4nt, 4-acetylbenzoic acid methyl ester, methyl4-acetylbenzoate, 4-acetyl-benzoic acid methyl ester, methyl 4-acetyl benzoate, pubchem22273, acmc-209ikx, #, ksc497k9b
PubChem CID 137990
IUPAC Name methyl 4-acetylbenzoate
SMILES CC(=O)C1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 93.0°C to 95.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3OOCC6H4COCH3
Quantity 1 g
Solubility Information Solubility in water: negligible
Formula Weight 178.19
Percent Purity 99+%
Physical Form Amorphous Powder
Chemical Name or Material Methyl 4-acetylbenzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.