missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, 99%, Thermo Scientific™

Product Code. 10562071 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10562071

Brand: Thermo Scientific Acros 292780010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 159660-85-2
Molecular Formula C14H18N2O2
Molecular Weight (g/mol) 246.31
MDL Number MFCD00216930
InChI Key HLKWMBMESNOAMS-UHFFFAOYSA-N
Synonym methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate, 1-benzyl-4-amino-1,2,5,6-tetrahydropyridine-3-carboxylic acid methyl ester, methyl 4-amino-1-benzyl-1,2,5,6-tetrahydro-3-pyridinecarboxylate #, methyl 4-amino-1-benzyl-5,6-dihydro-2h-pyridine-3-carboxylate, 3-pyridinecarboxylicacid, 4-amino-1,2,5,6-tetrahydro-1-phenylmethyl-, methyl ester, 4-amino-1-benzyl-1,2,5,6-tetrahydro-pyridine-3-carboxylic acid methyl ester, pyridine-3-carboxylic acid, 1,2,5,6-tetrahydro-4-amino-1-benzyl-, methyl ester
PubChem CID 563385
IUPAC Name methyl 4-amino-1-benzyl-3,6-dihydro-2H-pyridine-5-carboxylate
SMILES COC(=O)C1=C(CCN(C1)CC2=CC=CC=C2)N

Specifications

Melting Point 87.0°C to 89.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 246.31
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methyl 4-amino-1-benzyl-1,2,5,6-tetrahydropyridine-3-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.