missing translation for 'onlineSavingsMsg'
Learn More

methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride, Thermo Scientific™

Product Code. 10635155
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10635155 5 g 5g
10249412 1 g 1g
10487174 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10635155 Supplier Thermo Scientific Maybridge Supplier No. MO00998DE
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 180258-45-1
Molecular Formula C7H11ClN2O2
Molecular Weight (g/mol) 190.63
MDL Number MFCD01318761
InChI Key HEOKCJUUKIPIMM-UHFFFAOYSA-N
Synonym methyl 4-amino-1-methyl-1h-pyrrole-2-carboxylate hydrochloride, methyl 4-amino-1-methylpyrrole-2-carboxylate hydrochloride, methyl-4-amino-1-methyl pyrrole-2-carboxylate hydrochloride, 4-amino-1-methyl-1h-pyrrole-2-carboxylic acid-methyl ester hcl, 4-amino-1-methylpyrrole-2-carboxylic acid methyl ester hydrochloride, h-nh 4-mepyl-2-ome hcl, methyl 4-amino-1-methyl-pyrrole-2-carboxylate hydrochloride, methyl-4-amino-1-methyl pyrrole-2-carboxylate, hcl, n-methyl-2-carbomethoxy-4-aminopyrrole hydrochloride, 3-amino-1-methyl-5-methyloxycarbonylpyrrol hydrochloride
PubChem CID 2794656
SMILES Cl.COC(=O)C1=CC(N)=CN1C

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 5 g
IUPAC Name methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride
Formula Weight 190.63
Percent Purity 95%
Chemical Name or Material Methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.