missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-aminosalicylate, TRC

Product Code. 30478620
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478620 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478620 Supplier Toronto Research Chemicals Supplier No. TRCM328665500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 4-Amino-2-hydroxybenzoate
CAS 4136-97-4
Molecular Formula C8 H9 N O3
Quantity 500 mg
Synonym Methyl 4-Amino-2-hydroxybenzoate, 4-Amino-2-hydroxybenzoic acid methyl ester, Salicylic acid, 4-amino-, methyl ester (6CI,7CI,8CI), 2-Hydroxy-4-aminobenzoic acid methyl ester, 3-Hydroxy-4-methoxycarbonylaniline, 4-Amino-2-hydroxybenzoic acid methyl ester, 4-Aminosalicylic acid methyl ester, Methyl 2-hydroxy-4-aminobenzoate, Methyl 4-amino-2-hydroxybenzoate, Methyl 4-aminosalicylate, Methyl p-aminosalicylate, NSC 30263, NSC 48927, p-Aminosalicylic acid methyl ester
InChI Formula InChI=1S/C8H9NO3/c1-12-8(11)6-3-2-5(9)4-7(6)10/h2-4,10H,9H2,1H3
SMILES COC(=O)c1ccc(N)cc1O
IUPAC Name methyl 4-amino-2-hydroxybenzoate
Molecular Weight (g/mol) 167.16
Formula Weight 167.0582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.