missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Azido-4,6-dideoxy-2-O-methyl-3-O-benzyl-Alpha-D-glucopyranoside, TRC

Product Code. 30482327
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482327 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482327 Supplier Toronto Research Chemicals Supplier No. TRCM28874510MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 4-Azido-4,6-dideoxy-2-O-methyl-3-O-benzyl-a-D-glucopy
CAS 861819-28-5
Molecular Formula C15 H21 N3 O4
Quantity 10 mg
InChI Formula InChI=1S/C15H21N3O4/c1-10-12(17-18-16)13(14(19-2)15(20-3)22-10)21-9-11-7-5-4-6-8-11/h4-8,10,12-15H,9H2,1-3H3/t10-,12-,13+,14-,15+/m1/s1
SMILES CO[C@H]1O[C@H](C)[C@@H](N=[N+]=[N-])[C@H](OCc2ccccc2)[C@H]1OC
IUPAC Name (2S,3R,4S,5R,6R)-5-azido-2,3-dimethoxy-6-methyl-4-phenylmethoxyoxane
Molecular Weight (g/mol) 307.34
Formula Weight 307.1532
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.