missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromo-3-hydroxythiophene-2-carboxylate, 97%

Product Code. 11459726
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459726 10 g 10g
11449726 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459726 Supplier Thermo Scientific Alfa Aesar Supplier No. B20249.09
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 95201-93-7 | C6H5BrO3S | 237.067 g/mol

The synthesis of the thienopyranone scaffold began with the alkylation of methyl 4-bromo-3-hydroxythiophene-2-carboxylate with N-acetylmorpholine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The synthesis of the thienopyranone scaffold began with the alkylation of methyl 4-bromo-3-hydroxythiophene-2-carboxylate with N-acetylmorpholine.

Solubility
Sparingly Soluble in water 0.95 g/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95201-93-7
Molecular Formula C6H5BrO3S
Molecular Weight (g/mol) 237.067
MDL Number MFCD00052081
InChI Key UFTXASQYKJFRKI-UHFFFAOYSA-N
Synonym 2z-4-bromo-2-hydroxy methoxy methylidene thiophen-3-one, 2-thiophenecarboxylic acid, 4-bromo-3-hydroxy-, methyl ester, acmc-20akgk, maybridge1_004012, methyl 4-bromo-3-hydroxy-2-thiophenecarboxylate, methyl-3-hydroxy-4-bromo-2-thiophenecarboxylate, methyl-4-bromo-3-hydroxy-2-thiophenecarboxylate, 4-bromo-3-oxo-2-thienylidene-methoxy-methanolate, 4-bromo-2-hydroxy methoxy methylidene thiophen-3-one, 2-thiophenecarboxylicacid,4-bromo-3-hydroxy-,methyl ester
PubChem CID 2777611
IUPAC Name methyl 4-bromo-3-hydroxythiophene-2-carboxylate
SMILES COC(=O)C1=C(C(=CS1)Br)O

Specifications

Melting Point 79°C to 80°C
Quantity 10 g
Solubility Information Sparingly Soluble in water 0.95g/L @ 25°C.
Formula Weight 237.08
Percent Purity 97%
Chemical Name or Material Methyl 4-bromo-3-hydroxythiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.