missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromobutyrate, 98+%

Product Code. 10500922 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10500922 50 g Glass bottle 50g
10252522 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10500922 Supplier Thermo Scientific Acros Supplier No. 365830500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4897-84-1 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4897-84-1
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00041482
InChI Key QAWFLJGZSZIZHO-UHFFFAOYSA-N
Synonym methyl 4-bromobutyrate, 4-bromobutyric acid methyl ester, methyl-4-bromobutyrate, butanoic acid, 4-bromo-, methyl ester, methyl4-bromobutyrate, butyric acid, 4-bromo-, methyl ester, methyl gamma-bromobutyrate, acmc-1amn9, 4-brombutyric acid methyl ester, 4-bromobutyric acid methyl-ester
PubChem CID 107604
IUPAC Name methyl 4-bromobutanoate
SMILES COC(=O)CCCBr

Specifications

Color Colorless to Yellow
Density 1.4340g/mL
Boiling Point 186.0°C to 187.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4610 to 1.4630 (20°C, 589nm)
Quantity 50 g
Specific Gravity 1.434
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 181.03
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyl 4-bromobutyrate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-bromobutyrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.