missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromopyrrole-2-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15436807
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436807 5 g 5g
15426807 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436807 Supplier Thermo Scientific Alfa Aesar Supplier No. H33918.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 934-05-4
Molecular Formula C6H6BrNO2
Molecular Weight (g/mol) 204.023
MDL Number MFCD00832855
InChI Key ZXFCRVGOHJHZNF-UHFFFAOYSA-N
Synonym methyl 4-bromopyrrole-2-carboxylate, 4-bromo-2-methoxycarbonyl-1h-pyrrole, 4-bromo-1h-pyrrole-2-carboxylic acid methyl ester, methyl4-bromo-1h-pyrrole-2-carboxylate, 4-bromopyrrole-2-carboxylic acid methyl ester, 1h-pyrrole-2-carboxylic acid, 4-bromo-, methyl ester, pubchem13045, acmc-209rlm, ksc497g1p, 4-bromo-2-methoxycarbonylpyrrole
PubChem CID 2763594
IUPAC Name methyl 4-bromo-1H-pyrrole-2-carboxylate
SMILES COC(=O)C1=CC(=CN1)Br

Specifications

Melting Point 100°C to 102°C
Quantity 5 g
Formula Weight 204
Percent Purity 97%
Chemical Name or Material Methyl 4-bromopyrrole-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.