missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chlorobutyrate, 98%

Product Code. 11312286
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312286 100 g 100g
11302286 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312286 Supplier Thermo Scientific Alfa Aesar Supplier No. L03734.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3153-37-5 | C5H9ClO2 | 136.575 g/mol

Methyl 4-chlorobutyrate is used as a pharmaceutical synthesis, industrial dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-chlorobutyrate is used as a pharmaceutical synthesis, industrial dye intermediates.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3153-37-5
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.575
MDL Number MFCD00001003
InChI Key ZZUYIRISBMWFMV-UHFFFAOYSA-N
Synonym methyl 4-chlorobutyrate, butanoic acid, 4-chloro-, methyl ester, 4-chlorobutyric acid methyl ester, butyric acid, 4-chloro-, methyl ester, methyl .omega.-chlorobutyrate, 4-chlorobutanoic acid methyl ester, .gamma.-chlorobutyric acid methyl ester, methyl-4-chlorbutanoat, gamma-chlorobutyric acid methyl ester, methyl omega-chlorobutyrate
PubChem CID 76612
IUPAC Name methyl 4-chlorobutanoate
SMILES COC(=O)CCCCl

Specifications

Density 1.125
Boiling Point 175°C to 176°C
Flash Point 59°C (138°F)
Refractive Index 1.432
Quantity 100 g
UN Number UN3272
Beilstein 1745117
Solubility Information Slightly soluble in water.
Formula Weight 136.58
Percent Purity 98%
Chemical Name or Material Methyl 4-chlorobutyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.