missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chlorophenylacetate, 99%

Product Code. 10324352 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10324352 5 g Glass bottle 5g
10323612 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10324352 Supplier Thermo Scientific Acros Supplier No. 351050050
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52449-43-1 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52449-43-1
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00032743
InChI Key WWIYGBWRUXQDND-UHFFFAOYSA-N
Synonym methyl 4-chlorophenylacetate, methyl 2-4-chlorophenyl acetate, methyl 4-chlorophenyl acetate, 4-chloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 4-chloro-, methyl ester, 4-chlorophenylacetic acid methyl ester, methyl4-chlorophenylacetate, zlchem 382
PubChem CID 104196
IUPAC Name methyl 2-(4-chlorophenyl)acetate
SMILES COC(=O)CC1=CC=C(Cl)C=C1

Specifications

Color Yellow
Density 1.2000g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5220 to 1.5240
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.2
Linear Formula ClC6H4CH2CO2CH3
Beilstein 09, III, 2264
Formula Weight 184.62
Percent Purity 99%
Chemical Name or Material Methyl 4-chlorophenylacetate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 4-chlorophenylacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.