missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Chloropicolinate, TRC

Product Code. 30464713
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464713 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464713 Supplier Toronto Research Chemicals Supplier No. TRCM2944551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24484-93-3
Molecular Formula C7 H6 Cl N O2
Quantity 1 g
InChI Formula InChI=1S/C7H6ClNO2/c1-11-7(10)6-4-5(8)2-3-9-6/h2-4H,1H3
SMILES COC(=O)c1cc(Cl)ccn1
IUPAC Name methyl 4-chloropyridine-2-carboxylate
Molecular Weight (g/mol) 171.58
Formula Weight 171.0087
Chemical Name or Material Methyl 4-Chloropicolinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.