missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-ethynylbenzoate, 97%

Product Code. 15140126
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15140126 1 g 1g
15460208 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15140126 Supplier Thermo Scientific Alfa Aesar Supplier No. H53419.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3034-86-4 | C10H8O2 | 160.17 g/mol

Methyl 4-ethynylbenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-ethynylbenzoate is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water (0.16 g/L at 25°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-86-4
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00168820
InChI Key JPGRSTBIEYGVNO-UHFFFAOYSA-N
Synonym 4-ethynylbenzoic acid methyl ester, 4-ethynyl-benzoic acid methyl ester, benzoic acid, 4-ethynyl-, methyl ester, methyl4-ethynylbenzoate, 4-ethynylbenzoic acid methylester, 4-methoxycarbonyl phenylacetylene, benzoic acid,4-ethynyl-, methyl ester, methyl 4-ethynyl-benzoate, 4-ethinylbenzoic acid methyl ester, 4-ethynyl benzoic acid methylester
PubChem CID 640163
IUPAC Name methyl 4-ethynylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)C#C

Specifications

Melting Point 92°C to 94°C
Quantity 1 g
Solubility Information Sparingly soluble in water (0.16g/L at 25°C).
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material Methyl 4-ethynylbenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.