missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-formyl-1-methyl-1H-pyrrole-2-carboxylate, 95%, Thermo Scientific™

Product Code. 10703843
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10703843 1 g Amber Glass Bottle 1g
10203672 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10703843 Supplier Thermo Scientific Maybridge Supplier No. SEW06271DA
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 67858-47-3
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD01571196
InChI Key GXSZDESAIZEFGZ-UHFFFAOYSA-N
Synonym methyl 4-formyl-1-methyl-1h-pyrrole-2-carboxylate, pubchem12439, methylformylmethylpyrrolecarboxylate, 1h-pyrrole-2-carboxylicacid, 4-formyl-1-methyl-, methyl ester, 4-formyl-1-methyl-1h-pyrrole-2-carboxylic acid methyl ester
PubChem CID 2798349
IUPAC Name methyl 4-formyl-1-methylpyrrole-2-carboxylate
SMILES CN1C=C(C=C1C(=O)OC)C=O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 167.16
Percent Purity 95%
Chemical Name or Material Methyl 4-formyl-1-methyl-1H-pyrrole-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.