missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-formylbicyclo[2.2.2]octane-1-carboxylate, TRC

Product Code. 30479557
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479557 10 mg 10mg
30476795 50 mg 50mg
30473987 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479557 Supplier Toronto Research Chemicals Supplier No. TRCM30589010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94994-25-9
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.24
Synonym 4-Formyl-Bicyclo[2.2.2]Octane-1-Carboxylic Acid Methyl Ester
IUPAC Name methyl 1-formylbicyclo[2.2.2]octane-4-carboxylate
SMILES COC(=O)C12CCC(CC1)(CC2)C=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H16O3/c1-14-9(13)11-5-2-10(8-12,3-6-11)4-7-11/h8H,2-7H2,1H3
Formula Weight 196.1099
Chemical Name or Material Methyl 4-formylbicyclo[2.2.2]octane-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.