missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-methoxycarbonylbenzoylacetate, 95%, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsSpecifications
Specifications
| Melting Point | 81°C to 86°C |
| Color | White to Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min |
| Packaging | Glass bottle |
| Quantity | 1 g |
| Synonym | methyl 4-3-methoxy-3-oxopropanoyl benzoate, methyl 4-methoxycarbonylbenzoylacetate, 4-2-methoxycarbonyl-acetyl-benzoic acid methyl ester, 4-methoxycarbonyl-beta-oxobenzenepropanoic acid methyl ester, benzenepropanoic acid,4-methoxycarbonyl-a-oxo-, methyl ester, acmc-20efie, methyl 4-methoxycarbonyl benzoyl acetate, methyl4-methoxycarbonylbenzoylacetate, 4-methoxycarbonylbenzoyl acetic acid methyl ester, methyl 3-4-methoxycarbonyl phenyl-3-oxopropanoate |
| Solubility Information | Solubility in water: insoluble in water. Other solubilities: slightly soluble in acetone,methanol |
| Formula Weight | 236.22 |
| Percent Purity | 95% |
| Show More |
Safety and Handling
Product Identifier
- Methyl 4-methoxycarbonylbenzoylacetate
- Danger
- Acute toxicity Category 3
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?