missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxyphenylacetate, 97+%

Product Code. 11489066
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489066 25 g 25g
11499066 100 g 100g
11409076 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489066 Supplier Thermo Scientific Alfa Aesar Supplier No. A17940.14
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 23786-14-3 | C10H12O3 | 180.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23786-14-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00008454
InChI Key ZQYLDVNTWDEAJI-UHFFFAOYSA-N
Synonym methyl 4-methoxyphenylacetate, methyl 2-4-methoxyphenyl acetate, methyl 4-methoxyphenyl acetate, 4-methoxyphenylacetic acid methyl ester, 4-methoxy-phenyl-acetic acid methyl ester, methyl 4-methoxybenzeneacetate, methyl p-methoxyphenyl acetate, benzeneacetic acid, 4-methoxy-, methyl ester, acetic acid, p-methoxyphenyl-, methyl ester, methyl 2-p-methoxyphenyl acetate
PubChem CID 90266
IUPAC Name methyl 2-(4-methoxyphenyl)acetate
SMILES COC(=O)CC1=CC=C(OC)C=C1

Specifications

Density 1.135
Boiling Point 157°C to 159°C (19 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.516
Quantity 25 g
Beilstein 2209665
Formula Weight 180.23
Percent Purity ≥97%
Chemical Name or Material Methyl 4-methoxyphenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.