missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Methoxyphenylacetate, TRC

Product Code. 30468834
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30468834 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468834 Supplier Toronto Research Chemicals Supplier No. TRCM3384932.5G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 4-Methoxyphenylacetate
CAS 23786-14-3
Molecular Formula C10H12O3
Quantity 2.5 g
InChI Formula InChI=1S/C10H12O3/c1-12-9-5-3-8(4-6-9)7-10(11)13-2/h3-6H,7H2,1-2H3
SMILES COC(=O)Cc1ccc(OC)cc1
IUPAC Name methyl 2-(4-methoxyphenyl)acetate
Molecular Weight (g/mol) 180.2
Formula Weight 180.0786
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.