missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxypyridine-2-carboxylate, 98%

Product Code. 15461769
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
missing translation for 'unitSize':
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
15461769 25 g 25g
15441769 1 g 1g
15451769 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15461769 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' H64519.14
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29681-43-4 | C8H9NO3 | 167.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29681-43-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00956223
InChI Key OJDKENGKKYVJLY-UHFFFAOYSA-N
Synonym methyl 4-methoxypicolinate, 4-methoxy-pyridine-2-carboxylic acid methyl ester, 4-methoxy-pyridine-2-carboxylicacidmethylester, 2-pyridinecarboxylic acid, 4-methoxy-, methyl ester, 4-methoxypicolinic acid methyl ester, 4-methoxypyridine-2-carboxylic acid methyl ester, methyl 4-methoxy-2-pyridinecarboxylate, 4-methoxy-pyridine-2-carboxylic acid metyl ester, pubchem11083, acmc-1b7sb
PubChem CID 1519345
IUPAC Name methyl 4-methoxypyridine-2-carboxylate
SMILES COC1=CC(=NC=C1)C(=O)OC

Specifications

Melting Point 47°C to 50°C
Color White to Yellow
Quantity 25 g
Beilstein 473379
Formula Weight 167.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Methyl 4-methoxypyridine-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.