missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-oxopiperidine-3-carboxylate hydrochloride, 95%

Product Code. 11496440
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496440 5 g 5g
11362399 1 g 1g
11372399 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496440 Supplier Thermo Scientific Alfa Aesar Supplier No. H27318.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71486-53-8 | C7H12ClNO3 | 193.627 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71486-53-8
Molecular Formula C7H12ClNO3
Molecular Weight (g/mol) 193.627
MDL Number MFCD00012769
InChI Key NMAACQILAGCQPR-UHFFFAOYSA-N
Synonym methyl 4-oxopiperidine-3-carboxylate hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-methoxycarbonyl-4-piperidone hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hcl, 3-piperidinecarboxylic acid, 4-oxo-, methyl ester, hydrochloride, 3-methoxycarbonyl-4-oxopiperidinium chloride, methyl 4-oxopiperidine-3-carboxylate, chloride, 4-piperidine carboxylic acid methyl ketone 3-hydrochloride, pubchem12958, methyl 4-oxopiperidine-3-carboxylate hcl
PubChem CID 2724028
IUPAC Name methyl 4-oxopiperidine-3-carboxylate;hydrochloride
SMILES COC(=O)C1CNCCC1=O.Cl

Specifications

Melting Point ∽175°C (decomposition)
Quantity 5 g
Beilstein 4273241
Formula Weight 193.63
Percent Purity 95%
Chemical Name or Material Methyl 4-oxopiperidine-3-carboxylate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.