missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate, 95%, Thermo Scientific™

Product Code. 10407252
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10407252 10 g Amber Glass Bottle 10g
10572463 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10407252 Supplier Thermo Scientific Maybridge Supplier No. SEW02756EA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 237385-98-7
Molecular Formula C13H9F3O2S
Molecular Weight (g/mol) 286.268
MDL Number MFCD00110251
InChI Key RDMSDMSSVFMCKQ-UHFFFAOYSA-N
Synonym methyl 4-phenyl-5-trifluoromethyl thiophene-2-carboxylate, methyl 4-phenyl-5-trifluoromethyl-thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 4-phenyl-5-trifluoromethyl-, methyl ester, 2-methoxycarbonyl-4-phenyl-5-trifluoromethyl thiophene, methyl 4-phenyl-5-trifluoromethyl-2-thiophenecarboxylate, methyl 4-phenyl-5-trifluoromethyl thiophene-2carboxylate, 4-phenyl-5-trifluoromethyl-2-thiophenecarboxylic acid methyl ester, 4-phenyl-5-trifluoromethyl-thiophene-2-carboxylic acid methyl ester
PubChem CID 2781442
IUPAC Name methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
SMILES COC(=O)C1=CC(=C(S1)C(F)(F)F)C2=CC=CC=C2

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 286.27
Percent Purity 95%
Chemical Name or Material Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.