missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-pyrrolidin-1-ylbenzoate, 97%, Thermo Scientific™

Product Code. 10046332
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10046332 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10046332 Supplier Thermo Scientific Maybridge Supplier No. CC30122DA
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 129414-26-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H15NO2
MDL Number MFCD00731259
Quantity 1 g
Synonym methyl 4-pyrrolidin-1-yl benzoate, methyl 4-1-pyrrolidinyl benzenecarboxylate, 1-4-methoxycarbonyl phenyl pyrrolidine, 4-1-pyrrolidinyl benzoic acid methyl ester, benzoicacid, 4-1-pyrrolidinyl-, methyl ester, methyl 4-pyrrolidinylbenzoate, acmc-1bxvx, methyl 4-pyrrolidin-1-1ylbenzoate, methyl4-pyrrolidin-1-yl benzoate, 4-pyrrolidin-1-yl-benzoic acid methyl ester
InChI Key XNIVNLQKURGIAY-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=C(C=C1)N1CCCC1
IUPAC Name methyl 4-(pyrrolidin-1-yl)benzoate
Molecular Weight (g/mol) 205.26
PubChem CID 6971085
Formula Weight 205.26
Percent Purity 97%
Chemical Name or Material Methyl 4-pyrrolidin-1-ylbenzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.