missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(trifluoromethyl)benzoate, TRC

Product Code. 30478080
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478080 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478080 Supplier Toronto Research Chemicals Supplier No. TRCM332118500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2967-66-0
Molecular Formula C9H7F3O2
Quantity 500 mg
Synonym p-Toluic acid, alpha,alpha,alpha-trifluoro-, methyl ester (6CI,8CI), 4-Methoxycarbonylbenzotrifluoride, 4-Trifluoromethylbenzoic acid methyl ester, Methyl p-(trifluoromethyl)benzoate
InChI Formula InChI=1S/C9H7F3O2/c1-14-8(13)6-2-4-7(5-3-6)9(10,11)12/h2-5H,1H3
SMILES COC(=O)c1ccc(cc1)C(F)(F)F
IUPAC Name methyl 4-(trifluoromethyl)benzoate
Molecular Weight (g/mol) 204.15
Formula Weight 204.0398
Chemical Name or Material Methyl 4-(trifluoromethyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.