missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-acetylsalicylate, 98%, Thermo Scientific™

Product Code. 10491962 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10491962 5 g Glass Bottle 5g
10091801 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10491962 Supplier Thermo Scientific Acros Supplier No. 339180050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16475-90-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00040924
InChI Key XLSMGNNWSRNTIQ-UHFFFAOYSA-N
PubChem CID 85444
IUPAC Name methyl 5-acetyl-2-hydroxybenzoate
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)OC

Specifications

Melting Point 58°C to 62°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3C(O)C6H3(OH)COOCH3
Quantity 5 g
Synonym methyl 5-acetylsalicylate, 5-acetylsalicylic acid methyl ester, 5-acetyl-2-hydroxybenzoic acid methyl ester, methyl 2-hydroxy-5-acetylbenzoate, benzoic acid, 5-acetyl-2-hydroxy-, methyl ester, 5-acetyl methyl salicylate, methyl 5-acethylsalicylate, pubchem22504, methyl5-acetylsalicylate, methyl-5-acetylsalicylate
Formula Weight 194.19
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-acetylsalicylate, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.