missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-1-benzothiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10503403
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10503403 1 g Amber Glass Bottle 1g
10239932 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10503403 Supplier Thermo Scientific Maybridge Supplier No. MO00965DA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7312-11-0
Molecular Formula C10H7BrO2S
Molecular Weight (g/mol) 271.128
MDL Number MFCD05662034
InChI Key XDYVZHUZZZKQOS-UHFFFAOYSA-N
Synonym methyl 5-bromobenzo b thiophene-2-carboxylate, 5-bromo-benzo b thiophene-2-carboxylic acid methyl ester, benzo b thiophene-2-carboxylic acid, 5-bromo-, methyl ester, 5-bromo-benzo b thiophene-2-carboxylic methyl ester, maybridge4_002488, methyl 5-bromobenzo b thiophen-2-carboxylate, methyl 5-bromo-1-benzothiophene-2carboxylate, methyl 5-bromo-benzo b thiophene-2-carboxylate, methyl 5-bromobenzothiophene-2-carboxylate, 5-bromobenzothiophene-2-carboxylic acid methyl ester
PubChem CID 2824065
IUPAC Name methyl 5-bromo-1-benzothiophene-2-carboxylate
SMILES COC(=O)C1=CC2=C(S1)C=CC(=C2)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 271.13
Percent Purity 97%
Chemical Name or Material Methyl 5-bromo-1-benzothiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.