missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-2-furoate, 97%

Product Code. 11448887
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448887 5 g 5g
11438887 1 g 1g
11458887 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448887 Supplier Thermo Scientific Alfa Aesar Supplier No. B20524.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2527-99-3 | C6H5BrO3 | 205.007 g/mol

It is used in the preparation of hydroquinone bis(2-carbomethoxy-5-furyl ether).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of hydroquinone bis(2-carbomethoxy-5-furyl ether).

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2527-99-3
Molecular Formula C6H5BrO3
MDL Number MFCD00092313
InChI Key FBPIDMAELBIRLE-UHFFFAOYSA-N
Synonym methyl 5-bromo-2-furoate, methyl 5-bromo-2-furancarboxylate, 5-bromo-2-furoic acid methyl ester, 2-furancarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-furancarboxylic acid methyl ester, 5-bromofuran-2-carboxylic acid methyl ester, methyl 5-bromofuroate, 2-furoic acid, 5-bromo-, methyl ester, methyl 5-bromo-furanate, maybridge1_004996
PubChem CID 599633
IUPAC Name methyl 5-bromofuran-2-carboxylate

Specifications

Melting Point 62°C to 65°C
Quantity 5 g
Solubility Information Slightly soluble in water.
SMILES COC(=O)C1=CC=C(O1)Br
Molecular Weight (g/mol) 205.007
Formula Weight 205.01
Percent Purity 97%
Chemical Name or Material Methyl 5-bromo-2-furoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.