missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-2-(methylsulfanyl)-4-pyrimidinecarboxylate, 97%, Thermo Scientific™

Product Code. 10388813
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10388813 5 g Amber Glass Bottle 5g
10317644 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10388813 Supplier Thermo Scientific Maybridge Supplier No. MO01274DE
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 50593-91-4
Molecular Formula C7H7BrN2O2S
Molecular Weight (g/mol) 263.11
MDL Number MFCD05664428
InChI Key MYZJIEWTRJTWCD-UHFFFAOYSA-N
Synonym methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate, methyl 5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl pyrimidine-4-carboxylate, methyl 5-bromo-2-methylthio-4-pyrimidinecarboxylate, methylbromomethylsulfanylpyrimidinecarboxylate, methyl 5-bromo-2-methylthiopyrimidine-4-carboxylate, methyl-5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarb, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate,97, 5-bromo-2-methylsulfanylpyrimidine-4-carboxylic acid methyl ester
PubChem CID 2794729
SMILES COC(=O)C1=NC(SC)=NC=C1Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
IUPAC Name methyl 5-bromo-2-methylsulfanylpyrimidine-4-carboxylate
Formula Weight 263.11
Percent Purity 97%
Chemical Name or Material Methyl 5-bromo-2-(methylsulfanyl)-4-pyrimidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.