missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15448906
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448906 5 g 5g
15458906 250 mg 250mg
15438906 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448906 Supplier Thermo Scientific Alfa Aesar Supplier No. H64417.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

It can be used as pharmaceutical raw material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water (0.42 g/L).

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50593-91-4
Molecular Formula C7H7BrN2O2S
Molecular Weight (g/mol) 263.11
MDL Number MFCD05664428
InChI Key MYZJIEWTRJTWCD-UHFFFAOYSA-N
Synonym methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate, methyl 5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl pyrimidine-4-carboxylate, methyl 5-bromo-2-methylthio-4-pyrimidinecarboxylate, methylbromomethylsulfanylpyrimidinecarboxylate, methyl 5-bromo-2-methylthiopyrimidine-4-carboxylate, methyl-5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarb, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate,97, 5-bromo-2-methylsulfanylpyrimidine-4-carboxylic acid methyl ester
PubChem CID 2794729
SMILES COC(=O)C1=NC(SC)=NC=C1Br

Specifications

Melting Point 61°C to 64°C
Color Yellow
Quantity 5 g
UN Number UN2811
Beilstein 910703
Solubility Information Slightly soluble in water (0.42g/L).
IUPAC Name methyl 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylate
Formula Weight 263.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.