missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromofuran-2-carboxylate, TRC

Product Code. 30479573
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479573 100 mg 100mg
30480523 500 mg 500mg
30469100 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479573 Supplier Toronto Research Chemicals Supplier No. TRCM296140100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2527-99-3
Molecular Formula C6 H5 Br O3
Molecular Weight (g/mol) 205.006
IUPAC Name methyl 5-bromofuran-2-carboxylate
SMILES COC(=O)c1oc(Br)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H5BrO3/c1-9-6(8)4-2-3-5(7)10-4/h2-3H,1H3
Formula Weight 203.942
Chemical Name or Material Methyl 5-bromofuran-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.