missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromopyridine-2-carboxylate, 98+%, Thermo Scientific Chemicals

Product Code. 11457096
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11457096 5 g 5g
11447096 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11457096

Brand: Thermo Scientific Alfa Aesar B25415.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl 5-bromopyridine-2-carboxylate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-bromopyridine-2-carboxylate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29682-15-3
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD04112493
InChI Key JEURNBCYNWNADN-UHFFFAOYSA-N
Synonym methyl 5-bromopicolinate, 5-bromopyridine-2-carboxylic acid methyl ester, methyl 5-bromo-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyridinecarboxylic acid methyl ester, 5-bromo-pyridine-2-carboxylic acid methyl ester, methyl5-bromopicolinate, pubchem10589, 5-bromopyridine-2-carboxylicacidmethylester, acmc-1cjr7
PubChem CID 7016458
IUPAC Name methyl 5-bromopyridine-2-carboxylate
SMILES COC(=O)C1=NC=C(C=C1)Br

Specifications

Melting Point 99°C to 103°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 216.03
Percent Purity ≥98%
Chemical Name or Material Methyl 5-bromopyridine-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.