missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromovalerate, 97%

Product Code. 10606712 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 10606712

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5454-83-1 | C6H11BrO2 | 195.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5454-83-1
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000265
InChI Key RAVVJKCSZXAIQP-UHFFFAOYSA-N
PubChem CID 79557
IUPAC Name methyl 5-bromopentanoate
SMILES COC(=O)CCCCBr

Specifications

Color Brown to Colorless
Density 1.3600g/mL
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula Br(CH2)4CO2CH3
Refractive Index 1.4620 to 1.464
Quantity 25 g
Specific Gravity 1.36
Synonym methyl 5-bromovalerate, 5-bromovaleric acid methyl ester, pentanoic acid, 5-bromo-, methyl ester, methyl .omega.-bromopentanoate, 5-bromopentanoic acid methyl ester, 5-bromo-pentanoic acid methyl ester, methyl omega-bromopentanoate, methyl5-bromovalerate, valeric acid, 5-bromo-, methyl ester, methyl-5-bromovalerate
Formula Weight 195.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 5-bromovalerate, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.