missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-(chloromethyl)-2-furancarboxylate, 99%, Thermo Scientific™

Product Code. 10486052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10486052 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10486052 Supplier Thermo Scientific Acros Supplier No. 367410010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2144-37-8
Molecular Weight (g/mol) 174.58
MDL Number MFCD00020922
InChI Key PWXMEBZOKUPCST-UHFFFAOYSA-N
Synonym methyl 5-chloromethyl-2-furoate, methyl 5-chloromethyl furan-2-carboxylate, methyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-furan-2-carboxylic acid methyl ester, 2-furancarboxylic acid, 5-chloromethyl-, methyl ester, 5-chloromethylfuran-2-carboxylic acid methyl ester, pubchem24261, enamine_005466, acmc-1ck5d
PubChem CID 75065
IUPAC Name methyl 5-(chloromethyl)furan-2-carboxylate
SMILES COC(=O)C1=CC=C(O1)CCl

Specifications

Melting Point 31°C to 35°C
Color White to Yellow
Boiling Point 102.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7ClO3
Quantity 1 g
Formula Weight 174.58
Percent Purity 99%
Physical Form Oily Liquid or Low Melting Crystalline Solid
Chemical Name or Material Methyl 5-(chloromethyl)-2-furancarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 5-(chloromethyl)-2-furancarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.