missing translation for 'onlineSavingsMsg'
Learn More

Methyl-5-deoxy-2,3-O-isopropylidene-Beta-D-ribofuranoside, TRC

Product Code. 30474205
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474205 50 mg 50mg
30478442 100 mg 100mg
30467356 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474205 Supplier Toronto Research Chemicals Supplier No. TRCM82567050MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23202-81-5
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
Synonym Methyl-5-deoxy-2,3-O-(1-methylethylidene)-beta-D-ribofuranoside
IUPAC Name (3aR,4R,6R,6aR)-4-methoxy-2,2,6-trimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxole
SMILES CO[C@@H]1O[C@H](C)[C@H]2OC(C)(C)O[C@@H]12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H16O4/c1-5-6-7(8(10-4)11-5)13-9(2,3)12-6/h5-8H,1-4H3/t5-,6-,7-,8-/m1/s1
Formula Weight 188.1049
Chemical Name or Material Methyl-5-deoxy-2,3-O-isopropylidene-beta-D-ribofuranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.