missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-nitro-2-furoate, 97%

Product Code. 11462258
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462258 5 g 5g
11472258 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462258 Supplier Thermo Scientific Alfa Aesar Supplier No. A15226.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1874-23-3 | C6H5NO5 | 171.108 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1874-23-3
Molecular Formula C6H5NO5
Molecular Weight (g/mol) 171.108
MDL Number MFCD00051956
InChI Key UTLKCGPAJUYGOM-UHFFFAOYSA-N
Synonym methyl 5-nitro-2-furoate, methyl nitrofuroate, methyl 5-nitrofuroate, 5-nitro-2-furoic acid methyl ester, methyl 5-nitropyromucate, methyl 5-nitro-2-furancarboxylate, unii-71qm4e6j90, ccris 2152, 5-nitro-2-furancarboxylic acid methyl ester, 2-furancarboxylic acid, 5-nitro-, methyl ester
PubChem CID 94882
IUPAC Name methyl 5-nitrofuran-2-carboxylate
SMILES COC(=O)C1=CC=C(O1)[N+](=O)[O-]

Specifications

Melting Point 78°C to 82°C
Quantity 5 g
Beilstein 165132
Formula Weight 171.11
Percent Purity 97%
Chemical Name or Material Methyl 5-nitro-2-furoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.