missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-oxopyrrolidine-2-carboxylate, 95%, Thermo Scientific™

Product Code. 15462169
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462169 250 mg 250mg
15442169 1 g 1g
15452169 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15462169 Supplier Thermo Scientific Alfa Aesar Supplier No. H64804.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54571-66-3
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.14
MDL Number MFCD03844651
InChI Key HQGPKMSGXAUKHT-UHFFFAOYNA-N
Synonym methyl dl-pyroglutamate, 5-oxo-pyrrolidine-2-carboxylic acid methyl ester, methyl 5-oxo-2-pyrrolidinecarboxylate, 5-oxo-2-pyrrolidinecarboxylic acid methyl ester, methyl 5-oxo-dl-prolinate, methyl 5-oxo-2-pyrrolidinecarboxylate #, proline, 5-oxo-, methylester, 5-oxo-dl-proline methyl ester, methyl 2-pyrolidone-5-carboxylate, dl-pyroglutamic acid methyl ester
PubChem CID 500249
IUPAC Name methyl 5-oxopyrrolidine-2-carboxylate
SMILES COC(=O)C1CCC(=O)N1

Specifications

Melting Point 21°C to 23°C (literature)
Boiling Point 133°C to 135°C (1 mmHg)
Refractive Index 1.485
Quantity 250 mg
Beilstein 81950
Formula Weight 143.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 5-oxopyrrolidine-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.