missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Bromo-4-chloronicotinate, TRC

Product Code. 30482983
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482983 1 g 1g
30467033 250 mg 250mg
30470617 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482983 Supplier Toronto Research Chemicals Supplier No. TRCM2944831G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256789-73-7
Molecular Formula C7H5NO2ClBr
Molecular Weight (g/mol) 250.47
Synonym Methyl 6-Bromo-4-chloropyridine-3-carboxylate
IUPAC Name methyl 6-bromo-4-chloropyridine-3-carboxylate
SMILES COC(=O)c1cnc(Br)cc1Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5BrClNO2/c1-12-7(11)4-3-10-6(8)2-5(4)9/h2-3H,1H3
Formula Weight 248.9192
Chemical Name or Material Methyl 6-Bromo-4-chloronicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.